Search
Search

PISD| Gene Symbol: PISD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567539.2 | XP_005567596.1 | 7 - 8 | 1367 | 55 | |
| XM_005567540.2 | XP_005567597.1 | 7 - 8 | 1052 | 55 | ||
| XM_005567541.2 | XP_005567598.1 | 9 - 10 | 1428 | 55 | ||
| XM_005567542.2 | XP_005567599.1 | 9 - 10 | 1785 | 55 | ||
| XM_005567543.2 | XP_005567600.1 | 7 - 8 | 1194 | 55 | ||
| XM_005567544.2 | XP_005567601.1 | 7 - 8 | 1836 | 55 | ||
| XM_005567545.2 | XP_005567602.1 | 7 - 8 | 1207 | 55 | ||
| XM_005567547.2 | XP_005567604.1 | 7 - 8 | 1509 | 55 | ||
| XM_015457306.1 | XP_015312792.1 | 6 - 7 | 1080 | 55 | ||
| XM_015457307.1 | XP_015312793.1 | 8 - 9 | 1533 | 55 | ||
| XM_015457308.1 | XP_015312794.1 | 8 - 9 | 1611 | 55 | ||
| XM_015457309.1 | XP_015312795.1 | 7 - 8 | 1123 | 55 | ||
| XM_015457310.1 | XP_015312796.1 | 7 - 8 | 1476 | 55 | ||
| XM_015457311.1 | XP_015312797.1 | 7 - 8 | 1257 | 55 | ||
| XM_015457312.1 | XP_015312798.1 | 8 - 9 | 1262 | 55 | ||
Set Membership: |
|