Search
Search

GIMAP1| Gene Symbol: GIMAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551149.2 | XP_005551206.1 | 2 - 3 | 150 | 66 | |
| XM_005551150.2 | XP_005551207.1 | 2 - 3 | 177 | 66 | ||
Set Membership: |
|