Search
Search
| Gene Symbol: Bmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007556.2 | NP_031582.1 | 1 - 2 | 873 | 77 | |
| GenBank mRNA | AK041210.1 | - | 1231 | 77 | ||
| AK135770.1 | - | 1 - 2 | 506 | 77 | ||
| BC138593.1 | - | 1 - 2 | 793 | 8861613 | 77 | |
| BC138595.1 | - | 1 - 2 | 793 | 8861615 | 77 | |
| J04566.1 | - | 1 - 2 | 455 | 77 | ||
| X80992.1 | - | 1 - 2 | 872 | 77 | ||
Set Membership: |
|