Search
Search
| Gene Symbol: Fgd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008001.4 | NP_032027.2 | 11 - 12 | 2435 | 62 | |
| GenBank mRNA | AK148151.1 | - | 11 - 12 | 2436 | 62 | |
| BC004838.1 | - | 3 - 4 | 325 | 62 | ||
| BC011462.1 | - | 11 - 12 | 2372 | 3674293 | 62 | |
| U22325.1 | - | 11 - 12 | 2034 | 62 | ||
Set Membership: |
|