Search
Search

Itga7| Gene Symbol: Itga7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008398.2 | NP_032424.2 | 14 - 15 | 2175 | 58 | |
| GenBank mRNA | AK157612.1 | - | 14 - 15 | 2182 | 58 | |
| CX208029.1 | - | 1 - 2 | 41 | 58 | ||
| L23423.1 | - | 14 - 15 | 2173 | 58 | ||
Set Membership: |
|