Search
Search

Fxyd3| Gene Symbol: Fxyd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008557.2 | NP_032583.1 | 1 - 2 | 82 | 64 | |
| GenBank mRNA | AA959274.1 | - | 1 - 2 | 39 | 64 | |
| BC051033.1 | - | 1 - 2 | 40 | 4460667 | 64 | |
| BC056223.1 | - | 1 - 2 | 50 | 6485767 | 64 | |
| BG067532.2 | - | 7 - 8 | 64 | |||
| X93038.1 | - | 1 - 2 | 78 | 64 | ||
Set Membership: |
|