Search
Search
| Gene Symbol: Por | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC003240.1 | - | 3 - 4 | 336 | 3497749 | 68 |
| BC031463.1 | - | 3 - 4 | 281 | 3497979 | 68 | |
| D17571.1 | - | 3 - 4 | 280 | 68 | ||
Set Membership: |
|