Search
Search
| Gene Symbol: Ccr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009915.2 | NP_034045.1 | 2 - 3 | 124 | 100 | |
| GenBank mRNA | AK046579.1 | - | 358 | 100 | ||
| AK138599.1 | - | 1 - 2 | 437 | 100 | ||
| BC138803.1 | - | 1 - 2 | 84 | 8861825 | 100 | |
| BC138804.1 | - | 1 - 2 | 84 | 8861826 | 100 | |
| U51717.1 | - | 1 - 2 | 82 | 100 | ||
| U56819.1 | - | 2 - 3 | 124 | 100 | ||
Set Membership: |
|