Search
Search

Clcn5| Gene Symbol: Clcn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243762.1 | NP_001230691.1 | 8 - 9 | 1329 | 96 | |
| NM_016691.4 | NP_057900.3 | 6 - 7 | 947 | 96 | ||
| GenBank mRNA | AF134117.1 | - | 6 - 7 | 947 | 96 | |
| AK052990.1 | - | 6 - 7 | 927 | 96 | ||
| AK142227.1 | - | 6 - 7 | 878 | 96 | ||
| AK145763.1 | - | 6 - 7 | 916 | 96 | ||
| BC036347.1 | - | 6 - 7 | 919 | 3989812 | 96 | |
Set Membership: |
|