Search
Search
| Gene Symbol: Pld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143930.1 | - | 6 - 7 | 896 | 80 | |
| AK170701.1 | - | 6 - 7 | 888 | 80 | ||
| BC047268.1 | - | 6 - 7 | 820 | 80 | ||
| BC068317.1 | - | 6 - 7 | 833 | 30094606 | 80 | |
| U87557.1 | - | 6 - 7 | 698 | 80 | ||
Set Membership: |
|