Search
Search

Pold1| Gene Symbol: Pold1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011131.3 | NP_035261.3 | 9 - 10 | 1190 | 62 | |
| GenBank mRNA | AK088310.1 | - | 9 - 10 | 1181 | 62 | |
| AK146837.1 | - | 9 - 10 | 1186 | 62 | ||
| AK166592.1 | - | 9 - 10 | 1384 | 62 | ||
| AK167569.1 | - | 9 - 10 | 1173 | 62 | ||
| AK168967.1 | - | 9 - 10 | 1153 | 62 | ||
| AK169040.1 | - | 9 - 10 | 1186 | 62 | ||
| BC009128.1 | - | 9 - 10 | 1191 | 3257174 | 62 | |
| BC052670.1 | - | 8 - 9 | 1157 | 30058314 | 62 | |
| Z21848.1 | - | 9 - 10 | 1159 | 62 | ||
Set Membership: |
|