Search
Search
| Gene Symbol: Strn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011500.2 | NP_035630.2 | 16 - 17 | 2156 | 83 | |
| GenBank mRNA | AJ223777.1 | - | 16 - 17 | 2083 | 83 | |
| AK135684.1 | - | 11 - 12 | 1349 | 83 | ||
| AK136032.1 | - | 14 - 15 | 1805 | 83 | ||
| BC090968.1 | - | 16 - 17 | 2156 | 30646517 | 83 | |
| BC150727.1 | - | 16 - 17 | 2124 | 9087638 | 83 | |
Set Membership: |
|