Search
Search

Casc3| Gene Symbol: Casc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138660.2 | NP_619601.2 | 9 - 10 | 1693 | 83 | |
| GenBank mRNA | AF526276.1 | - | 9 - 10 | 1667 | 83 | |
| AJ292072.1 | - | 1907 | 83 | |||
| AK037399.1 | - | 3 - 4 | 288 | 83 | ||
| AK139608.1 | - | 9 - 10 | 1732 | 83 | ||
| AK145886.1 | - | 9 - 10 | 1467 | 83 | ||
| BC034533.1 | - | 6 - 7 | 1337 | 83 | ||
| BC060672.1 | - | 9 - 10 | 1521 | 83 | ||
| BC141296.1 | - | 9 - 10 | 1806 | 9055908 | 83 | |
Set Membership: |
|