Search
Search
| Gene Symbol: Crot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023733.3 | NP_076222.1 | 11 - 12 | 1222 | 95 | |
| GenBank mRNA | AK004567.2 | - | 11 - 12 | 1223 | 95 | |
| BC006593.1 | - | 11 - 12 | 1210 | 3156501 | 95 | |
| BC012308.1 | - | 11 - 12 | 1237 | 3500167 | 95 | |
Set Membership: |
|