Search
Search
| Gene Symbol: Dpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031843.2 | NP_114031.2 | 6 - 7 | 736 | 75 | |
| GenBank mRNA | AF285235.1 | - | 6 - 7 | 731 | 75 | |
| AK028293.1 | - | 4 - 5 | 1357 | 75 | ||
| AK140406.1 | - | 5 - 6 | 850 | 75 | ||
| AK140420.1 | - | 5 - 6 | 850 | 75 | ||
| AK154193.1 | - | 6 - 7 | 746 | 75 | ||
| AK211757.1 | - | 1 - 2 | 41 | 75 | ||
| BC027205.1 | - | 6 - 7 | 736 | 4190473 | 75 | |
| BC052861.1 | - | 4 - 5 | 1364 | 75 | ||
Set Membership: |
|