Search
Search

Steap4| Gene Symbol: Steap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054098.3 | NP_473439.2 | 2 - 3 | 527 | 62 | |
| GenBank mRNA | AJ319746.1 | - | 2 - 3 | 530 | 62 | |
| AK040760.1 | - | 2 - 3 | 543 | 62 | ||
| BC006651.1 | - | 2 - 3 | 527 | 3500056 | 62 | |
Set Membership: |
|