Search
Search

Elmo2| Gene Symbol: Elmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080287.2 | NP_525026.2 | 17 - 18 | 1789 | 93 | |
| NM_207706.1 | NP_997589.1 | 18 - 19 | 1825 | 93 | ||
| GenBank mRNA | AB093301.1 | - | 8 - 9 | 2370 | 93 | |
| AF398884.1 | - | 16 - 17 | 1688 | 93 | ||
| AK032033.1 | - | 18 - 19 | 1798 | 93 | ||
| AK033918.1 | - | 17 - 18 | 1983 | 93 | ||
| AK035710.1 | - | 2 - 3 | 175 | 93 | ||
| AK038097.1 | - | 17 - 18 | 1778 | 93 | ||
| AK038455.1 | - | 17 - 18 | 1895 | 93 | ||
| AK045428.1 | - | 17 - 18 | 1790 | 93 | ||
| AK047040.1 | - | 17 - 18 | 1751 | 93 | ||
| AK050823.1 | - | 17 - 18 | 1752 | 93 | ||
| AK053574.1 | - | 18 - 19 | 1789 | 93 | ||
| AK129023.1 | - | 17 - 18 | 1789 | 93 | ||
| BC006678.1 | - | 3 - 4 | 392 | 93 | ||
| BC023954.1 | - | 18 - 19 | 1810 | 5320429 | 93 | |
| BC033418.1 | - | 16 - 17 | 1719 | 93 | ||
Set Membership: |
|