Search
Search

Npdc1| Gene Symbol: Npdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008721.4 | NP_032747.2 | 6 - 7 | 933 | 54 | |
| GenBank mRNA | AK146466.1 | - | 6 - 7 | 822 | 54 | |
| BC003320.1 | - | 6 - 7 | 900 | 3155595 | 54 | |
| BC023887.1 | - | 5 - 6 | 967 | 5151868 | 54 | |
| L03814.1 | - | 6 - 7 | 737 | 54 | ||
| X67209.1 | - | 6 - 7 | 737 | 54 | ||
Set Membership: |
|