Search
Search
| Gene Symbol: Pts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011220.2 | NP_035350.1 | 2 - 3 | 171 | 85 | |
| GenBank mRNA | AF043225.1 | - | 2 - 3 | 186 | 85 | |
| AK002614.1 | - | 2 - 3 | 181 | 85 | ||
| AK146987.1 | - | 2 - 3 | 182 | 85 | ||
| BC029013.1 | - | 2 - 3 | 171 | 5357873 | 85 | |
Set Membership: |
|