Search
Search

Pcsk1| Gene Symbol: Pcsk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013628.2 | NP_038656.1 | 6 - 7 | 780 | 103 | |
| GenBank mRNA | BC108982.1 | - | 6 - 7 | 758 | 40047652 | 103 |
| BC108983.1 | - | 6 - 7 | 758 | 40047655 | 103 | |
| M55668.1 | - | 1 - 2 | 66 | 103 | ||
| M58589.1 | - | 6 - 7 | 780 | 103 | ||
| M69196.1 | - | 6 - 7 | 886 | 103 | ||
Set Membership: |
|