Search
Search

Rilpl1| Gene Symbol: Rilpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021430.2 | NP_067405.1 | 5 - 6 | 1387 | 97 | |
| GenBank mRNA | AB041584.1 | - | 5 - 6 | 1017 | 97 | |
| AK013469.1 | - | 2 - 3 | 280 | 97 | ||
| BC029965.1 | - | 5 - 6 | 1065 | 97 | ||
| BC051945.1 | - | 5 - 6 | 1387 | 5716540 | 97 | |
| BC051946.1 | - | 5 - 6 | 1317 | 6403995 | 97 | |
| BC058384.1 | - | 5 - 6 | 1280 | 5718399 | 97 | |
Set Membership: |
|