Search
Search

Ints6| Gene Symbol: Ints6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008715.2 | NP_032741.2 | 15 - 16 | 2551 | 95 | |
| GenBank mRNA | BC059263.1 | - | 15 - 16 | 2551 | 6409580 | 95 |
Set Membership: |
|