Search
Search

Trim26| Gene Symbol: Trim26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025599.3 | NP_001020770.2 | 3 - 4 | 701 | 69 | |
| NM_001286726.1 | NP_001273655.1 | 3 - 4 | 720 | 69 | ||
| NM_001286727.1 | NP_001273656.1 | 3 - 4 | 669 | 69 | ||
| GenBank mRNA | AK042252.1 | - | 3 - 4 | 684 | 69 | |
| AK148247.1 | - | 3 - 4 | 607 | 69 | ||
| AK171019.1 | - | 3 - 4 | 674 | 69 | ||
| BC037110.1 | - | 3 - 4 | 683 | 5369645 | 69 | |
Set Membership: |
|