Search
Search

Spata5| Gene Symbol: Spata5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163511.1 | NP_001156983.1 | 13 - 14 | 2281 | 60 | |
| NM_021343.2 | NP_067318.2 | 13 - 14 | 2278 | 60 | ||
| GenBank mRNA | AF049099.1 | - | 13 - 14 | 2260 | 60 | |
| AK011111.2 | - | 13 - 14 | 2245 | 60 | ||
| AK144998.1 | - | 13 - 14 | 2608 | 60 | ||
| BC145302.1 | - | 13 - 14 | 2249 | 9053856 | 60 | |
| BC145309.1 | - | 13 - 14 | 2249 | 60 | ||
Set Membership: |
|