| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2436438 |
BC108362.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2436438 |
BC080718.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2436438 |
AK149363.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2436438 |
AK148708.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2436438 |
NM_001037955.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2071092 |
BC057944.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2095213 |
BC108362.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2095213 |
BC080718.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2095213 |
AK149363.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2095213 |
AK148708.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2095213 |
AF237619.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
128431 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776811 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
599682 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
22439 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
128795 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
74907 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
490846 |
AK003818.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
490846 |
AK005296.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
490846 |
AK011159.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
490846 |
AK030567.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
490846 |
AK161187.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
490846 |
BC027160.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
490846 |
NM_025442.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
215258 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
686427 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
186780 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
292886 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
320127 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
106812 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
143271 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
274863 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
286597 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
365142 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
304075 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
730282 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
501131 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
728776 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
746056 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
524646 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
367453 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
186343 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
453441 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
646521 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
5310 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
587259 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
630910 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
642282 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
791624 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
523573 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
151299 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
47124 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
149099 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
129681 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2095213 |
BC108362.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2095213 |
BC080718.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2095213 |
AK149363.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2095213 |
AK148708.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2095213 |
AF237619.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2436438 |
BC108362.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2436438 |
BC080718.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2436438 |
AK149363.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2436438 |
AK148708.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2436438 |
NM_001037955.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P231793 |
BC108362.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P231793 |
BC080718.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P231793 |
AK149363.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P231793 |
AK148708.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P231793 |
BC057944.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P231793 |
BC108362.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P231793 |
BC080718.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P231793 |
AK149363.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P231793 |
AK148708.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P231793 |
BC057944.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P378211 |
AF237619.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P378211 |
NM_134068.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P378211 |
AK003947.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P129552 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P137115 |
- |
- |