Search
Search

Calcoco1| Gene Symbol: Calcoco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026192.3 | NP_080468.1 | 8 - 9 | 1162 | 81 | |
| GenBank mRNA | AK007393.1 | - | 8 - 9 | 1053 | 81 | |
| AK018734.2 | - | 8 - 9 | 1048 | 81 | ||
| AK035598.1 | - | 8 - 9 | 1074 | 81 | ||
| AK087513.1 | - | 8 - 9 | 1116 | 81 | ||
| AK220400.1 | - | 8 - 9 | 1075 | 81 | ||
| AY131329.1 | - | 7 - 8 | 1028 | 81 | ||
| BC054783.1 | - | 8 - 9 | 1056 | 6815943 | 81 | |
Set Membership: |
|