| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P459012 |
BC120837.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P459012 |
EU233996.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P459012 |
NM_027964.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P459012 |
AK006367.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P459012 |
BC120837.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P459012 |
EU233996.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P459012 |
NM_027964.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P459012 |
AK006367.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354622 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
604184 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
45012 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
362364 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
102014 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
667830 |
AK013596.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
667830 |
BC066797.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
667830 |
BC089303.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
667830 |
NM_001081426.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809126 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
150686 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
12269 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
539037 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
450594 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
684131 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
55899 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P147689 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P143949 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P459012 |
BC120837.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P459012 |
EU233996.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P459012 |
NM_027964.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P459012 |
AK006367.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P459012 |
BC120837.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P459012 |
EU233996.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P459012 |
NM_027964.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P459012 |
AK006367.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P459012 |
BC120837.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P459012 |
EU233996.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P459012 |
NM_027964.2 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P459012 |
AK006367.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P459012 |
BC120837.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P459012 |
EU233996.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P459012 |
NM_027964.2 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P459012 |
AK006367.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390261 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390289 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390290 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390299 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390303 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390269 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390285 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390262 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390298 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390288 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390264 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390283 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390301 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390281 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390292 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390273 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390304 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390265 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390296 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390260 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390258 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390293 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390279 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390270 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390276 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390306 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390272 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390267 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390256 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390282 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21390277 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04589579 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P143949 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P147689 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P459012 |
BC120837.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P459012 |
EU233996.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P459012 |
NM_027964.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P459012 |
AK006367.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P459012 |
BC120837.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P459012 |
EU233996.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P459012 |
NM_027964.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P459012 |
AK006367.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P106259 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P134007 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P143949 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P147689 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01701137 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01597602 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01597599 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01701138 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01701143 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01597603 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390226 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390219 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390158 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390177 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390272 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390240 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390146 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390183 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390166 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390298 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390232 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390273 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390225 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390215 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390156 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390243 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390144 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390222 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390141 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390163 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390276 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390197 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390261 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390258 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390181 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390191 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390281 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390211 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390290 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390180 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390301 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390250 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390154 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390254 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390151 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390147 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390207 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390260 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390194 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390199 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390229 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390230 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390179 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390223 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390285 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390184 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390262 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390224 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390198 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390303 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390235 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390153 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390170 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390288 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390256 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390139 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390304 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390282 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390292 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390217 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390269 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390265 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390296 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390193 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390161 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390214 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390267 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390142 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390228 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390167 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390252 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390148 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390204 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390306 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390264 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390152 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390279 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390202 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390160 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390242 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390178 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21390299 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01629990 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390283 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390303 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390254 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390214 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390291 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390290 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390219 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390220 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390228 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390211 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02047996 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390285 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390224 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390226 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390267 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390288 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390279 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390304 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390276 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01293231 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01173236 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390225 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390294 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01903653 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390282 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390261 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390301 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390252 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390272 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390277 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390258 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390260 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390273 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390281 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390250 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390222 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390306 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01293224 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390262 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390215 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390217 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390213 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390278 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390289 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01665473 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390296 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390223 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390256 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390265 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21390299 |
- |
- |