Search
Search

Agap3| Gene Symbol: Agap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139153.2 | NP_631892.2 | 11 - 12 | 1525 | 82 | |
| GenBank mRNA | AF459091.1 | - | 11 - 12 | 1525 | 82 | |
| AK079516.1 | - | 7 - 8 | 864 | 82 | ||
Set Membership: |
|