Search
Search

Slc24a4| Gene Symbol: Slc24a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172152.2 | NP_742164.1 | 6 - 7 | 553 | 85 | |
| GenBank mRNA | AK044239.1 | - | 6 - 7 | 1242 | 85 | |
| AK044368.1 | - | 6 - 7 | 1246 | 85 | ||
| AY156046.1 | - | 6 - 7 | 553 | 85 | ||
| BC137945.1 | - | 6 - 7 | 581 | 8860964 | 85 | |
| BC171939.1 | - | 6 - 7 | 581 | 9054225 | 85 | |
Set Membership: |
|