| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605618 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605725 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605622 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605617 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605728 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605619 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605723 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605616 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605727 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605724 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1997977 |
BC061513.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1997977 |
BI732997.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1997977 |
BC046334.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1997977 |
BC054719.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1997977 |
AK171898.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
327908 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
679727 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
541633 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
465976 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
939384 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
824279 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
339125 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
567082 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
740678 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
421150 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
248871 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
315937 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
AK003179.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
AK005202.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
AK008394.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
AK014665.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
AK029950.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
AK046088.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
AK086937.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
AK164299.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
BC016565.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
BC050116.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
BC098490.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
NM_001110129.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
NM_001110130.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
NM_028677.4 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
XM_485828.3 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
XM_905767.2 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
XM_980747.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
XM_983649.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
XM_990471.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897209 |
XM_991392.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
952090 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
299761 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
56292 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
82152 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
3255 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P134984 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P132891 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P101907 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1997977 |
BC061513.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1997977 |
BI732997.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1997977 |
BC046334.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1997977 |
BC054719.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1997977 |
AK171898.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P107936 |
BC061513.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P107936 |
BC046334.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P107936 |
BC054719.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P107936 |
BC033312.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P107936 |
AK169657.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P446132 |
BC061513.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P446132 |
BI732997.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P446132 |
BC046334.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P446132 |
BC054719.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P446132 |
AK171898.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605716 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605706 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605714 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605708 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605728 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605720 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605704 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605725 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605711 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605719 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605713 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605700 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605727 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605705 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605723 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605701 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605717 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605709 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29605722 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P132891 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08112669 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P134984 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P101907 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02850627 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P107936 |
BC061513.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P107936 |
BC046334.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P107936 |
BC054719.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P107936 |
BC033312.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P107936 |
AK169657.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P446132 |
BC061513.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P446132 |
BI732997.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P446132 |
BC046334.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P446132 |
BC054719.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P446132 |
AK171898.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P109598 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P134984 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P101907 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P132891 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11835549 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11588533 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11835556 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605616 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605617 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605618 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605724 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605723 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605619 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605622 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605725 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605727 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29605728 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605745 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605648 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605640 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605651 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605755 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605605 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605666 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605760 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605606 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605759 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605626 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605642 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605727 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605752 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605637 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605730 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605597 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605735 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605595 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605674 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605619 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605660 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605620 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605776 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605601 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605653 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605769 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605617 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605748 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605614 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605615 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605628 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605725 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605607 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605599 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605633 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605604 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605645 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605676 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605654 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605611 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605728 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605664 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605656 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605733 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605616 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605655 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605646 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605768 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605668 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605731 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605603 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605770 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605662 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605747 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605782 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605591 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605741 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605624 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605639 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605622 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605643 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605634 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605629 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29605596 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605757 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605616 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11306208 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605777 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605741 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605770 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11757196 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11178583 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11697521 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11178580 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605664 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605750 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605659 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605745 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11178594 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605727 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605626 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605666 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605738 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605748 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605766 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605663 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605759 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605608 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605611 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605762 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11178579 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11835555 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605622 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605746 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11474167 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605731 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605656 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11778953 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605725 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605733 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605609 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605624 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605665 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605760 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11754022 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605735 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605730 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605614 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605755 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605768 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605769 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605747 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605660 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605615 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605736 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605776 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605668 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29605618 |
- |
- |