| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700010531.mm.1 |
NM_001159672.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010531.mm.1 |
NM_001159672.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010531.mm.1 |
NM_175317.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010531.mm.1 |
NM_175317.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010531.mm.1 |
TC0700001187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700010530.mm.1 |
NM_001159672.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010530.mm.1 |
NM_001159672.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010530.mm.1 |
NM_175317.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010530.mm.1 |
NM_175317.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010530.mm.1 |
TC0700001187.mm.1 |
MTA TC |