Search
Search

Zmiz2| Gene Symbol: Zmiz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005867.1 | NP_001005867.1 | 17 - 18 | 2542 | 122 | |
| NM_028601.3 | NP_082877.2 | 17 - 18 | 2638 | 122 | ||
| GenBank mRNA | AK048533.1 | - | 532 | 122 | ||
| BC024081.1 | - | 16 - 17 | 2388 | 122 | ||
| BC034541.1 | - | 3 - 4 | 3131 | 122 | ||
| BC040745.1 | - | 8 - 9 | 1097 | 122 | ||
| BC060652.1 | - | 17 - 18 | 2480 | 30354393 | 122 | |
| BC063069.1 | - | 9 - 10 | 1243 | 122 | ||
Set Membership: |
|