Search
Search
| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012919.1 | - | 1 - 2 | 165 | 129 | |
| AK054412.1 | - | 1 - 2 | 154 | 129 | ||
| AK133177.1 | - | 1 - 2 | 163 | 129 | ||
| AK154594.1 | - | 1 - 2 | 172 | 129 | ||
| AK164249.1 | - | 1 - 2 | 163 | 129 | ||
| AK164999.1 | - | 1 - 2 | 170 | 129 | ||
| AK166351.1 | - | 1 - 2 | 172 | 129 | ||
| AK169601.1 | - | 1 - 2 | 163 | 129 | ||
| BB841951.1 | - | 1 - 2 | 193 | 129 | ||
| BC060208.1 | - | 1 - 2 | 124 | 129 | ||
| CF898098.1 | - | 1 - 2 | 186 | 129 | ||
Set Membership: |
|