Search
Search
| Gene Symbol: Ik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011879.2 | NP_036009.1 | 16 - 17 | 1524 | 89 | |
| GenBank mRNA | AJ006130.1 | - | 16 - 17 | 1407 | 89 | |
| AK082356.1 | - | 16 - 17 | 1456 | 89 | ||
| AK087976.1 | - | 16 - 17 | 1470 | 89 | ||
| BC006706.1 | - | 14 - 15 | 1882 | 89 | ||
| BC014739.1 | - | 16 - 17 | 1455 | 4920184 | 89 | |
| BC026569.1 | - | 16 - 17 | 1455 | 4982612 | 89 | |
| BC029148.1 | - | 14 - 15 | 2418 | 89 | ||
| BC029671.1 | - | 16 - 17 | 1467 | 5364953 | 89 | |
| BC029694.1 | - | 16 - 17 | 1464 | 5369477 | 89 | |
| BC033356.1 | - | 16 - 17 | 1451 | 5366230 | 89 | |
Set Membership: |
|