Search
Search

Ttll12| Gene Symbol: Ttll12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183017.2 | NP_898838.2 | 10 - 11 | 1531 | 67 | |
| GenBank mRNA | AK084879.1 | - | 10 - 11 | 1498 | 67 | |
| AK134345.1 | - | 10 - 11 | 1498 | 67 | ||
| AK143496.1 | - | 9 - 10 | 1364 | 67 | ||
| AK150116.1 | - | 10 - 11 | 1497 | 67 | ||
| AM690756.1 | - | 10 - 11 | 1484 | 67 | ||
| BC055368.1 | - | 10 - 11 | 1496 | 30055801 | 67 | |
Set Membership: |
|