Search
Search

Srebf1| Gene Symbol: Srebf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011480.3 | NP_035610.1 | 12 - 13 | 2412 | 86 | |
| GenBank mRNA | AF374266.1 | - | 11 - 12 | 2176 | 86 | |
| AK052628.1 | - | 12 - 13 | 2413 | 86 | ||
| AK154424.1 | - | 12 - 13 | 2380 | 86 | ||
| AK158269.1 | - | 5 - 6 | 961 | 86 | ||
| AK169607.1 | - | 12 - 13 | 2413 | 86 | ||
| BC006051.1 | - | 9 - 10 | 1548 | 86 | ||
| BC056922.1 | - | 12 - 13 | 2394 | 6824948 | 86 | |
Set Membership: |
|