Search
Search
| Gene Symbol: Tex9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009359.4 | NP_033385.2 | 10 - 11 | 952 | 148 | |
| GenBank mRNA | AK008505.1 | - | 10 - 11 | 950 | 148 | |
| AK018568.1 | - | 867 | 148 | |||
| AK032490.1 | - | 9 - 10 | 1060 | 148 | ||
| BC006667.1 | - | 1 - 2 | 1600 | 148 | ||
| BC031504.1 | - | 1 - 2 | 835 | 148 | ||
| BC141033.1 | - | 10 - 11 | 1094 | 9055645 | 148 | |
| BC145184.1 | - | 11 - 12 | 1193 | 148 | ||
| BC145185.1 | - | 10 - 11 | 1094 | 9053736 | 148 | |
| BC145186.1 | - | 9 - 10 | 920 | 9053737 | 148 | |
Set Membership: |
|