Search
Search

Kdm3b| Gene Symbol: Kdm3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081256.1 | NP_001074725.1 | 11 - 12 | 3326 | 81 | |
| GenBank mRNA | AK129281.1 | - | 10 - 11 | 2682 | 81 | |
| BC038376.1 | - | 1 - 2 | 144 | 81 | ||
| BC060727.1 | - | 4 - 5 | 886 | 81 | ||
| BC108415.1 | - | 4 - 5 | 730 | 81 | ||
| BC151084.1 | - | 11 - 12 | 3287 | 9087997 | 81 | |
| BC172150.1 | - | 10 - 11 | 2687 | 81 | ||
Set Membership: |
|