Search
Search

Apbb3| Gene Symbol: Apbb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146085.1 | NP_666197.1 | 8 - 9 | 1064 | 66 | |
| GenBank mRNA | AK141878.1 | - | 8 - 9 | 966 | 66 | |
| AK171634.1 | - | 7 - 8 | 1257 | 66 | ||
| AK210931.1 | - | 1 - 2 | 39 | 66 | ||
| BC024136.1 | - | 7 - 8 | 1062 | 66 | ||
| BC024809.1 | - | 8 - 9 | 1064 | 5357681 | 66 | |
Set Membership: |
|