Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159925.1 | NP_001153397.1 | 5 - 6 | 699 | 82 | |
NM_001159926.1 | NP_001153398.1 | 5 - 6 | 699 | 82 | ||
NM_011038.2 | NP_035168.1 | 5 - 6 | 699 | 82 | ||
GenBank mRNA | AB008912.1 | - | 5 - 6 | 674 | 82 | |
AB010557.1 | - | 5 - 6 | 642 | 82 | ||
AB010558.1 | - | 5 - 6 | 657 | 82 | ||
AF031150.1 | - | 5 - 6 | 732 | 82 | ||
AF104231.1 | - | 4 - 5 | 454 | 82 | ||
AF145233.1 | - | 5 - 6 | 646 | 82 | ||
BC131906.1 | - | 5 - 6 | 610 | 40130183 | 82 | |
BC131908.1 | - | 5 - 6 | 608 | 40130185 | 82 | |
BC145168.1 | - | 5 - 6 | 610 | 9053719 | 82 | |
BC145169.1 | - | 5 - 6 | 610 | 9053720 | 82 | |
Y09584.1 | - | 3 - 4 | 338 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |