Search
Search
| Gene Symbol: Has1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008215.2 | NP_032241.1 | 2 - 3 | 754 | 117 | |
| GenBank mRNA | BC125424.1 | - | 2 - 3 | 733 | 40129939 | 117 |
| BC125426.1 | - | 2 - 3 | 733 | 40129941 | 117 | |
| D82964.1 | - | 2 - 3 | 760 | 117 | ||
Set Membership: |
|