Search
Search

Fdx1l| Gene Symbol: Fdx1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039824.2 | NP_001034913.1 | 2 - 3 | 181 | 105 | |
| GenBank mRNA | AK008401.2 | - | 2 - 3 | 179 | 105 | |
| AK010211.1 | - | 2 - 3 | 152 | 105 | ||
| AK020979.1 | - | 2 - 3 | 185 | 105 | ||
| BC061189.1 | - | 1 - 2 | 268 | 175 | ||
| BC172154.1 | - | 1 - 2 | 351 | 175 | ||
Set Membership: |
|