| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900020057.mm.1 |
NM_001161535.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020057.mm.1 |
NM_001161535.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020057.mm.1 |
NM_001161537.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020057.mm.1 |
NM_001161537.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020057.mm.1 |
TC0900002388.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020058.mm.1 |
NM_001161535.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020058.mm.1 |
NM_001161535.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020058.mm.1 |
NM_001161537.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020058.mm.1 |
NM_001161537.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020058.mm.1 |
NM_001161540.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020058.mm.1 |
NM_001161540.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020058.mm.1 |
TC0900002388.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020059.mm.1 |
NM_001161540.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020059.mm.1 |
NM_001161540.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020059.mm.1 |
TC0900002388.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161535.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161535.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161536.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161536.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161537.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161537.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161538.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161538.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161539.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161539.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161540.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161540.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161541.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_001161541.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_177193.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
NM_177193.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020053.mm.1 |
TC0900002388.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020061.mm.1 |
NM_001161536.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020061.mm.1 |
NM_001161536.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020061.mm.1 |
NM_177193.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020061.mm.1 |
NM_177193.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020061.mm.1 |
TC0900002388.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020060.mm.1 |
NM_001161536.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020060.mm.1 |
NM_001161536.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020060.mm.1 |
NM_177193.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020060.mm.1 |
NM_177193.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020060.mm.1 |
TC0900002388.mm.1 |
MTA TC |