Search
Search

Tmprss13| Gene Symbol: Tmprss13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013373.2 | NP_001013391.2 | 3 - 4 | 91 | ||
| GenBank mRNA | BC042878.1 | - | 2 - 3 | 267 | 91 | |
| BC085323.1 | - | 3 - 4 | 516 | 30734093 | 91 | |
Set Membership: |
|