Search
Search

Lactb| Gene Symbol: Lactb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030717.1 | NP_109642.1 | 3 - 4 | 704 | 159 | |
| GenBank mRNA | AF317900.1 | - | 3 - 4 | 704 | 159 | |
| AK078550.1 | - | 3 - 4 | 645 | 159 | ||
| AK149410.1 | - | 3 - 4 | 647 | 159 | ||
| BC016583.1 | - | 3 - 4 | 631 | 159 | ||
| BC021604.1 | - | 3 - 4 | 630 | 159 | ||
| BC046293.1 | - | 3 - 4 | 631 | 6484767 | 159 | |
| BC147809.1 | - | 3 - 4 | 649 | 9087526 | 159 | |
Set Membership: |
|