Search
Search
| Gene Symbol: Bms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194339.1 | NP_919320.1 | 11 - 12 | 2263 | 90 | |
| GenBank mRNA | AK129082.1 | - | 11 - 12 | 2223 | 90 | |
| AK133013.1 | - | 11 - 12 | 2213 | 90 | ||
| AK134315.1 | - | 11 - 12 | 2271 | 90 | ||
| AK135031.1 | - | 10 - 11 | 2760 | 90 | ||
| AK162178.1 | - | 11 - 12 | 2321 | 90 | ||
| BC030906.1 | - | 4 - 5 | 1063 | 90 | ||
| BC057054.1 | - | 11 - 12 | 2263 | 6827225 | 90 | |
Set Membership: |
|