Search
Search

Nr2e1| Gene Symbol: Nr2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152229.2 | NP_689415.1 | 1 - 2 | 708 | 69 | |
| GenBank mRNA | AK132201.1 | - | 1 - 2 | 712 | 69 | |
| BC056950.1 | - | 1 - 2 | 339 | 69 | ||
| BC057104.1 | - | 1 - 2 | 752 | 6401770 | 69 | |
| S77482.1 | - | 1 - 2 | 751 | 69 | ||
Set Membership: |
|