| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400026888.mm.1 |
NR_024078.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026888.mm.1 |
NR_024078.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026888.mm.1 |
TC0400003360.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026890.mm.1 |
NR_024078.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026890.mm.1 |
NR_024078.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026890.mm.1 |
TC0400003360.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026879.mm.1 |
NR_024078.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026879.mm.1 |
NR_024078.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026879.mm.1 |
TC0400003360.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026886.mm.1 |
NR_024078.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026886.mm.1 |
NR_024078.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026886.mm.1 |
TC0400003360.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026883.mm.1 |
NR_024078.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026883.mm.1 |
NR_024078.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026883.mm.1 |
TC0400003360.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026889.mm.1 |
NR_024078.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026889.mm.1 |
NR_024078.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026889.mm.1 |
TC0400003360.mm.1 |
MTA TC |