Search
Search

B4galnt3| Gene Symbol: B4galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198884.1 | NP_942585.1 | 16 - 17 | 2493 | 66 | |
| GenBank mRNA | AB114826.1 | - | 16 - 17 | 2493 | 66 | |
| AK082874.1 | - | 2 - 3 | 211 | 66 | ||
| AK143898.1 | - | 16 - 17 | 2409 | 66 | ||
| BC139798.1 | - | 16 - 17 | 2507 | 40060840 | 66 | |
Set Membership: |
|