Search
Search
| Gene Symbol: Pnck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199351.1 | NP_001186280.1 | 3 - 4 | 388 | 68 | |
| NM_001199352.1 | NP_001186281.1 | 3 - 4 | 252 | 68 | ||
| NM_012040.3 | NP_036170.1 | 3 - 4 | 328 | 68 | ||
| GenBank mRNA | AB023027.1 | - | 2 - 3 | 198 | 68 | |
| AF181984.1 | - | 3 - 4 | 302 | 68 | ||
| AK134665.1 | - | 3 - 4 | 259 | 68 | ||
| AK158755.1 | - | 3 - 4 | 253 | 68 | ||
| BC051996.1 | - | 3 - 4 | 235 | 68 | ||
| BC055891.1 | - | 3 - 4 | 228 | 4208888 | 68 | |
| CX243772.1 | - | 3 - 4 | 388 | 68 | ||
Set Membership: |
|